1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt for spectrophotometric determination of Co, Fe, K, Ag, Ba, Ca, Co, Fe, Ni, Pb, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
3-Hydroxy-4-nitroso-2,7-naphthalenedisulphonic acid disodium salt, Nitroso-R salt
73950-100G
73950-25G
BJ73950-100GEA
197
USD
BJ73950-100G
BJ73950-25G
1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt for spectrophotometric determination of Co, Fe, K, Ag, Ba, Ca, Co, Fe, Ni, Pb, Fluka™
1-Nitroso-2-naphthol-3,6-disulphonic acid disodium salt
Formula:
C₁₀H₅NNa₂O₈S₂ Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003885 CAS Number: 525-05-3 EINECS: 208-369-1 |
Specification Test Results
Identity (IR) | complying |
Assay | 90.0 - 105.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...