Streptavidin, PE (Phycoerythrin), BioLegend
Supplier: Biolegend
405203 405204
405203-BLEA 81.3 USD
405203-BL 405204-BL
Streptavidin, PE (Phycoerythrin), BioLegend
Proteins and Peptides
Streptavidin binds to biotin with high affinity. Streptavidin-Phycoerythrin (PE) is used for detecting biotinylated antibodies. The excitation of PE by 488 nm laser light induces a light emission maximum of 575 nm.