Buffer Solutions, according to DIN 19266, Fluka™, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
33595-6X1L
82454-500ML
33594-6X1L
BJ33594-1LCS
415
USD
BJ33594-1L
BJ33595-1L
BJ82454-500ML
BJ33594-6X1L
Buffer Solutions, according to DIN 19266, Fluka™, Honeywell Research Chemicals
Buffers
pH Calibration Buffers
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...