Forceps, Stainless Steel, Sharp, Straight, Bürkle
Supplier: Bürkle
5386-0202 5386-0200 5386-0204
76319-778EA 9.25 USD
76319-778 76319-776 76319-774
Forceps, Stainless Steel, Sharp, Straight, Bürkle
Forceps
The sturdy forceps are ideal for use in laboratories.
  • Stainless steel AISI 304 (1.4301)
  • Flutings at the ends and grip for secure handling
Pointed ends with a fluted internal structure ensure reliable handling of different samples. The fluted handle offers a good grip. These laboratory tweezers are suitable for use in precise work in chemistry, the pharmaceutical industry, cosmetics, and food and beverage.