Forceps, Stainless Steel, Blunt, Straight, Bürkle
Supplier: BURKLE INC
5386-0300 5386-0302 5386-0304
76319-780EA 7.27 USD
76319-780 76319-850 76319-852
Forceps, Stainless Steel, Blunt, Straight, Bürkle
Forceps
These forceps are suitable for precise laboratory tasks.
  • Stainless steel AISI 304 (1.4301)
  • Flutings at the ends and grip for secure handling
These forceps have a straight shape with blunt ends that are fluted on the inside. This makes it possible to pick up different materials, tissues, or samples accurately.