Coomassie® brilliant blue R-250 solution

Supplier: BIO-RAD LABORATORIES MS
Danger

1610436 1610437
101643-252EA 116.92 USD
101643-252 101643-254
Coomassie® brilliant blue R-250 solution
Coomassie® brilliant blue R-250

Use this Coomassie brilliant blue R-250 solution to stain proteins in SDS-PAGE gels.


  • Ready to use for fast and easy staining
  • Mixture of water, methanol, and glacial acetic acid

Formula: C₄₅H₄₄N₃NaO₇S₂
Melting Pt: 174…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00041762
CAS Number: 6104-59-2
EINECS: 228-060-5
UN: 2924
ADR: 3 (8),III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR