Bio-Safe™ Coomassie Stain, Bio-Rad
Supplier: BIO-RAD LABORATORIES MS
1610786 1610787
101643-422EA 178.46 USD
101643-422 101643-424
Bio-Safe™ Coomassie Stain, Bio-Rad
Stains Electrophoresis Stains
Premixed staining solution, for polyacrylamide protein gels. Choose the flexibility of Bio-Safe Coomassie Premixed Staining Solution. Proteins can be visualized in polyacrylamide gels with or without the use of acetic acid/methanol fixation.
  • No requirement for fixation* with acetic acid and methanol
  • Destain with water
  • No handling and disposal of solvents
  • Gels can be stored in water
* Fixation is recommended for peptide and very low molecular weight protein separations prior to using Bio-Safe Coomassie Stain.
Sensitivity: ≥3 ng per band (QC Colloidal Coomassie); ≥8 ng per band (Bio-Safe™ Coomassie)