ChemMax® 1 Disposable Chemical Protective Boot Covers, Serged Seams and Elastic Top, Lakeland Industries
Supplier: Lakeland
C1S903YP-2X C1S903YP-XL
76314-348CS 462.76 USD
76314-348 76314-350 76314-352
ChemMax® 1 Disposable Chemical Protective Boot Covers, Serged Seams and Elastic Top, Lakeland Industries
Disposable Garments
These disposable chemical protective boot covers are used for protection from paint, hazardous liquids, chemical splash, and bloodborne pathogens.
  • Serged seams
  • Feature elastic top closures for a better fit and increased protection
These garments are manufactured from ChemMax® 1 polyethylene barrier film and continuous filament polypropylene, non-woven.