Culture Inserts, Ibidi
Supplier: IBIDI USA MS
80369 80209
MSPP-80369EA 384.62 USD
MSPP-80369 MSPP-80209
Culture Inserts, Ibidi
Culture Dishes Cell Culture Dishes
Culture inserts made of biocompatible silicone material with an adhesive underside.
  • Complete solution for wound healing experiments, requiring only a few steps from sample preparation to image analysis
  • Reproducible experiments owing to a defined 500 µm cell free gap, no leakage during cultivation, and no material being left behind after the insert’s removal
  • Suitable for doing experiments in parallel
  • Used for self-insertion into 6 or 12 well plates, or other formats
  • Cell free gap width of 500 µm +/- 100 µm
Suitable for wound healing, migration assays, 2D invasion assays, and co-cultivation of cells.
Envase: Pack of 25 pieces in a transport dish.