Thin Walled 'P' Series NMR Sample Tubes, 5 mm, New Era™
Supplier: New ERA Enterprises
NELP57 NEMP58 NEUP57 NEUP58 NESP58 NESP57 NELP58 NEHP57 NEMP57 NEHP585PK NEHP575PK
100165-882EA 13.28 USD
100165-882 100165-884 100165-944 100165-964 100165-976 100165-980 100165-880 100165-938 100165-886 103549-600 103549-602
Thin Walled 'P' Series NMR Sample Tubes, 5 mm, New Era™
Tubes NMR Tubes
These sample tubes are useful for air-moisture sensitive samples, organometallics, kinetics, and other samples requiring carefully controlled environments.
  • Made of type 1, class A borosilicate glass (Pyrex or equivalent)
  • Round bottom design
  • Each tube is capped and marked with the catalog number
Suitable for all experiments when matched to the application and field strength.