NMR Sample Tubes, 3 mm, New Era™
Supplier: New ERA Enterprises
NEH38 NEH37
100165-952EA 15.46 USD
100165-952 100165-936
NMR Sample Tubes, 3 mm, New Era™
Tubes NMR Tubes
Improved design allows for increased sample volume while maintaining adequate wall thickness to help reduce breakage. For use in all 3 mm micro probes.
  • Made of type 1, class A borosilicate glass
  • Tubes are suitable for flame sealing to Pyrex® or equivalent
  • Each tube is capped and marked with the catalog number
Ideal for biological samples, hands-on and high throughput/autosampler use, and samples not requiring special handling. Suitable for all experiments when matched to the application and instrument field strength.