2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: 2,2'-Azobis[2-(2-imidazolin-2-yl)propane] Dihydrochloride, 1,2-Bis(2-(4,5-dihydro-1H-imidazol-2-yl)propan-2-yl)diazene dihydrochloride, 2-{2-[(E)-2-[2-(4,5-Dihydro-1H-imidazol-2-yl)proan-2-yl]diazen-1-yl]propan-2-yl}-4,5-dihydro-1H-imidazole dihydrochloride

092374-25G 092374-100G
103607-322EA 43.08 USD
103607-322 103607-324
2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride ≥95%
2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride

Other name: 1,2-Bis(2-(4,5-dihydro-1H-imidazol-2-yl)-propan-2-yl)diazene dihydrochloride

Formula: C₁₂H₂₄Cl₂N₆
MW: 323.27 g/mol
MDL Number: MFCD00142723
CAS Number: 27776-21-2

Order Now

Specification Test Results

Purity ≥95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR