Cryo-Crystallography Magnetic Pin Tools, Rigaku
Supplier: Rigaku Reagents
1013197 1013198 1013199
103729-048PK 180.46 USD
103729-048 103729-050 103729-060
Cryo-Crystallography Magnetic Pin Tools, Rigaku
Magnets
Cryo-crystallography is an invaluable technique for single crystal X-ray diffraction and provides many benefits including mitigation of radiation damage.
  • Stainless steel tool with plunger release for manipulating magnetic pins in liquid nitrogen
  • Kit contains both straight and curved Crystal Wand
  • Straight and curved models are also available individually
The technique requires special tools and practical considerations to be successful. Rigaku Reagents is pleased to offer a number of these tools for cryo-crystallography.
Order Now
SPECIFICATIONS
Environmentally Preferable