Compact 300 Crystallization Plates (XJR), Rigaku
Supplier: Rigaku Reagents
1009545 1009546
103727-740CS 142.54 USD
103727-740 103727-742
Compact 300 Crystallization Plates (XJR), Rigaku
Microplates Crystallisation Plates
Compact 300 Crystallization plates utilize a patented sitting drop well design and vapor diffusion channel. These plates feature 96 reservoirs with 96 drop chambers.
  • Reservoir volume is 300 microliters
  • Drop chamber volume is 6 microliters
  • Plate dimensions: 85.5×127.8×14.4 mm
These crytallization plates are compatible with SBS (Society for Biomolecular Screening) standard laboratory automated instrumentation.
Drop chambers are 9 mm apart in both horizontal and vertical directions for eay use with multi-channel pipettors.