CombiClover 1500 Crystallization Plates (CBT), Rigaku
Supplier: Rigaku Reagents
1009543 1009544
103727-736CS 142.54 USD
103727-736 103727-738
CombiClover 1500 Crystallization Plates (CBT), Rigaku
Microplates Crystallisation Plates
CombiClover 1500 Crystallization plates offer the exact same design and utility as the CombiClover 500 crystallization plates, but offer larger capacity in the reservoir and drop chambers.
  • Central reservoir volume is 1.5 millilters
  • Satellite drop chamber volume is 50 microliters
  • Plate dimensions: 112×165×24 mm
A central reservoir is connected to four satellite drop chambers via dedicated vapor diffusion channels to create a novel combinatorial 'crystallization clover'.