CombiClover 500 Crystallization Plates (CJR), Rigaku
Supplier: Rigaku Reagents
1009541 1009542
103727-732CS 142.54 USD
103727-732 103727-734
CombiClover 500 Crystallization Plates (CJR), Rigaku
Microplates Crystallisation Plates
CombiClover 500 Crystallization Plates utilze a patented sitting-drop well design. A central reservoir is connected to four satellite drop chambers via dedicated vapor diffusion channels to create a novel combinatorial 'crystallization clover'.
  • Central reservoir volume is 500 microliters
  • Satellite drop chamber volume is 10 microliters
  • Plate dimensions: 85.5×127.8×14.4 mm
Each plate has 24 of the described clovers, providing 96 flat-bottom sitting drop chambers. The unique design allows four different crystallization experiments to occur within each clover, providing maximum efficacy and conservation of crystallization solutions.
Drop chambers are 9 mm apart and reservoir chambers are 18 mm apart in both horizontal and vertical directions for easy use with multi-channel pipettors.
These plates are compatible with SBS (Society for Biomolecular Screening) standard laboratory automated instrumentation.