Luer Lock Syringes, B. Braun
Supplier: B Braun
4617100V-02 4610303-02 4617207V-02
10753-946CS 514.7 USD
10753-946 76313-872 76313-876
Luer Lock Syringes, B. Braun
Syringes General-Purpose Syringes
These syringes provide consistent, dependable performance. They have a PP barrel and plunger rod and a polyisoprene plunger tip.
  • Smooth plunger action and double-seal stopper
  • High visibility- transparent barrel with bold graduations
  • Secure backstop to prevent accidental plunger withdrawal
  • Double seal stopper to prevent medication leakage
  • Large, stable finger grips
Components do not contain DEHP, PVC, or rubber tree latex.
Sterilization method: EO.
Total shelf life: 60 months.