Heparin sodium salt (from porcine intestinal mucosa), Calbiochem®

Supplier: MilliporeSigma
Warning

Synonyms: Reviparin sodium

375095-500KU 375095-100KU
80603-436EA 509.37 USD
80603-436 80058-186
Heparin sodium salt (from porcine intestinal mucosa), Calbiochem®
Heparin sodium salt

Preservative-free, non-sterile. Highly sulfated dextrorotatory mucopolysaccharide with specific anticoagulant properties. Inhibits the IP3-activated Ca²⁺ channel release of the endoplasmic reticulum.


  • Activity ≥160 USP units/mg dry weight
  • Form Lyophilized
  • M.W. 13,500-15,000 (Note: 1 KU = 1000 units)

Storage Temperature: Ambient
MDL Number: MFCD00081689
CAS Number: 9041-08-1
Merck Index: 14,4653

Order Now

Specification Test Results

Endotoxin ≤0.5 EU/mg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR