Ethanol denatured ≥70% (w/v) for histology, Supelco®
Supplier: THOMAS SCIENTIFIC INC MX
|
Danger
|
Denatured Alcohol (Proprietary Solvent) 70 volumes / Water 30 volumes
Denaturant: Exact percentages are being withheld as a trade secret.
methanol (>= 1 % - < 5 % )
4-methylpentan-2-one (>= 1 % - < 5 % )
ethyl acetate (>= 1 % - < 5 % )
Formula:
H₃CCH₂OH Density: 0.8750…0.8850 g/mL (20 °C) Flash Pt: 10.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 UN: 1993 ADR: 3,II |
Specification Test Results
Color | Colorless |
Denatured alcohol content | 68-72% by vol |
Density at 20�C | 0.8750-0.8850g/mL |
Form | Clear liquid |
Water content | 28-32% by vol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...