Vacuum Manifolds, Stainless Steel, Advantec
Supplier: Advantec MFS
351130 361120 353130 363130 351120 361130 313200 363330 353300 313000 363320 363120 353100 353330
101104-444PK 1235.23 USD
101104-444 101104-446 101103-872 101103-876 101104-442 101104-448 101104-510 101104-998 101104-992 101103-378 101104-996 101103-874 101103-870 101104-994
Vacuum Manifolds, Stainless Steel, Advantec
Manifolds
Manifolds are used under closed conditions and must be vented to atmosphere without breaking the vacuum connection. 3-way valve vent is female Luer slip, accepts standard disposable syringe filter or stainless steel syringe filter holder.
  • Sterility test manifold features rear mounted flushing manifold for use in USP bacteria retentive method
  • Exterior taper accepts a 40/35 female joint
  • Standard cup accepts a #8 stopper to adapt microanalysis units and other devices