Anti-REN Sheep Polyclonal Antibody

Supplier: Innovative Research Inc
SASRRENPSHT.1 SASRRENPSHT1MG
103386-148EA 375.38 USD
103386-148 103387-826
Anti-REN Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: REN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR