Anti-REN1 Sheep Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
SASRRENHTBIO.1 SASRRENHTBIO1
103386-144EA 436.92 USD
103386-144 103387-822
Anti-REN1 Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: REN1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR