Anti-PLAT Sheep Polyclonal Antibody

Supplier: Innovative Research Inc
SASRBTPA-1ML SASRBTPA-10ML
103386-112EA 375.38 USD
103386-112 103387-790
Anti-PLAT Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PLAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR