Anti-SERPINA10 Sheep Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
SASMZPIBIO10MG SASMZPIBIO1MG
103387-782EA 3483.08 USD
103387-782 103388-236
Anti-SERPINA10 Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: SERPINA10
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR