Anti-PLAU Sheep Polyclonal Antibody

Supplier: Innovative Research Inc
SASMUPA-GF-1MG SASMUPAGF10MG
103386-098EA 276.92 USD
103386-098 103387-760
Anti-PLAU Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PLAU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR