Anti-PLAT Sheep Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
SASMTPABIO1MG SASMTPABIO10MG
103386-076EA 375.38 USD
103386-076 103387-738
Anti-PLAT Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: PLAT
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR