Anti-SERPINE2 Sheep Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
SASMPN1BIO1MG SASMPN1BIO10MG
103386-052EA 436.92 USD
103386-052 103387-714
Anti-SERPINE2 Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: SERPINE2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR