Anti-SERPINI1 Sheep Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
SASMNSPBIO1MG SASMNSPBIO10MG
103386-028EA 436.92 USD
103386-028 103388-800
Anti-SERPINI1 Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: SERPINI1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR