Anti-F10 Sheep Polyclonal Antibody

Supplier: Innovative Research Inc
SASMFXHT0.1MG SASMFXGFHT1MG
103386-020EA 412.31 USD
103386-020 103388-462
Anti-F10 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: F10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR