Anti-F3 Sheep Polyclonal Antibody

Supplier: Innovative Research Inc
SASHTF-GF-5MG SASHTF-GF-1MG
103387-662EA 562.69 USD
103387-662 103388-220
Anti-F3 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR