Anti-CPB2 Sheep Polyclonal Antibody

Supplier: Innovative Research Inc
SASHTAFIGF5MG SASHTAFIGF1MG
103387-660EA 562.69 USD
103387-660 103388-218
Anti-CPB2 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CPB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR