Anti-REN Sheep Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
SASHRENHTBIO.1 SASHRENHTBIO1
103385-970EA 498.46 USD
103385-970 103387-656
Anti-REN Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: REN
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR