Anti-PROS1 Sheep Polyclonal Antibody

Supplier: Innovative Research Inc
SASHPS-GF-1MG SASHPS-GF-5MG
103385-944EA 255.77 USD
103385-944 103387-630
Anti-PROS1 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PROS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR