Anti-KLK3 Sheep Polyclonal Antibody

Supplier: Innovative Research Inc
SASHPSA-1ML SASHPSA-10ML
103385-932EA 276.92 USD
103385-932 103387-618
Anti-KLK3 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KLK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR