Anti-PLG Sheep Polyclonal Antibody

Supplier: Innovative Research Inc
SASHPLGHT0.1MG SASHPLGGFHT1MG
103385-928EA 276.92 USD
103385-928 103387-614
Anti-PLG Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PLG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR