Anti-PROC Sheep Polyclonal Antibody

Supplier: Innovative Research Inc
SASHPC-GF-1MG SASHPC-GF-5MG
103385-916EA 255.77 USD
103385-916 103387-602
Anti-PROC Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PROC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR