Anti-SERPINE1 Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Innovative Research Inc
SASHPAIHRP1MG SASHPAIHRP10MG
103385-912EA 344.62 USD
103385-912 103387-598
Anti-SERPINE1 Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Primary
Antigen: SERPINE1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR