Anti-F7 Sheep Polyclonal Antibody

Supplier: Innovative Research Inc
SASHFVIIGF1MG SASHFVIIGF5MG
103385-884EA 255.77 USD
103385-884 103388-792
Anti-F7 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: F7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR