Anti-TNNT2 Sheep Polyclonal Antibody

Supplier: Innovative Research Inc
SASHCTNTGF1MG SASHCTNTGF10MG
103385-864EA 276.92 USD
103385-864 103388-442
Anti-TNNT2 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TNNT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR