Anti-SERPIND1 Sheep Polyclonal Antibody

Supplier: Innovative Research Inc
SASHCII-GF-1MG SASHCII-GF-5MG
103385-846EA 255.77 USD
103385-846 103388-770
Anti-SERPIND1 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SERPIND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR