Anti-IgG Rabbit Polyclonal Antibody

Supplier: Innovative Research Inc
RBGFED1000MG RB-GF-ED-50MG
103387-462EA 2516.92 USD
103387-462 103388-188
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR