Anti-IgG Mouse Polyclonal Antibody

Supplier: Innovative Research Inc
MSBC-GF-10MG MSBC-GF-50MG
103385-500EA 221.54 USD
103385-500 103387-250
Anti-IgG Mouse Polyclonal Antibody
Antibodies
Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR