Anti-F2 Rat Monoclonal Antibody [clone: 9013]

Supplier: Innovative Research Inc
MA-MPT-0.1MG MA-MPT-0.5MG
103388-116EA 358.08 USD
103388-116 103388-378
Anti-F2 Rat Monoclonal Antibody [clone: 9013]
Antibodies
Type: Primary
Antigen: F2
Clonality: Monoclonal
Clone: 9013
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR