Anti-F10 Rat Monoclonal Antibody [clone: 9051]

Supplier: Innovative Research Inc
MA-MFX-2-0.1MG MA-MFX-2-0.5MG
103388-108EA 358.08 USD
103388-108 103388-370
Anti-F10 Rat Monoclonal Antibody [clone: 9051]
Antibodies
Type: Primary
Antigen: F10
Clonality: Monoclonal
Clone: 9051
Conjugation: Unconjugated
Epitope: Heavy Chain
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR