Anti-F8 Rat Monoclonal Antibody [clone: 9035]

Supplier: Innovative Research Inc
MAMFVIII0.5MG MAMFVIII0.1MG
103388-366EA 708.85 USD
103388-366 103388-862
Anti-F8 Rat Monoclonal Antibody [clone: 9035]
Antibodies
Type: Primary
Antigen: F8
Clonality: Monoclonal
Clone: 9035
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR