Anti-F2 Mouse Monoclonal Antibody [clone: 5020]

Supplier: Innovative Research Inc
MAHTHROM0.1MG MAHTHROM0.5MG
103385-412EA 321.54 USD
103385-412 103388-358
Anti-F2 Mouse Monoclonal Antibody [clone: 5020]
Antibodies
Type: Primary
Antigen: F2
Clonality: Monoclonal
Clone: 5020
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR