Anti-PROS1 Mouse Monoclonal Antibody [clone: 5092]

Supplier: Innovative Research Inc
MA-HPS-2-0.5MG MA-HPS-2-0.1MG
103387-140EA 643.08 USD
103387-140 103388-098
Anti-PROS1 Mouse Monoclonal Antibody [clone: 5092]
Antibodies
Type: Primary
Antigen: PROS1
Clonality: Monoclonal
Clone: 5092
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR