Anti-PROC Mouse Monoclonal Antibody [clone: 5072]

Supplier: Innovative Research Inc
MA-HPC-2-0.5MG MA-HPC-2-0.1MG
103387-138EA 643.08 USD
103387-138 103388-096
Anti-PROC Mouse Monoclonal Antibody [clone: 5072]
Antibodies
Type: Primary
Antigen: PROC
Clonality: Monoclonal
Clone: 5072
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR