Anti-LRP1 Mouse Monoclonal Antibody [clone: 8G1] (Biotin)

Supplier: Innovative Research Inc
MA-8G1-BIO-1MG MA8G1BIO0.1MG
103387-128EA 2270.77 USD
103387-128 103388-086
Anti-LRP1 Mouse Monoclonal Antibody [clone: 8G1] (Biotin)
Antibodies
Type: Primary
Antigen: LRP1
Clonality: Monoclonal
Clone: 8G1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR