Anti-LRP1 Mouse Monoclonal Antibody [clone: 8B8]

Supplier: Innovative Research Inc
MA-8B8-10MG MA-8B8-1MG
103387-120EA 4947.69 USD
103387-120 103388-078
Anti-LRP1 Mouse Monoclonal Antibody [clone: 8B8]
Antibodies
Type: Primary
Antigen: LRP1
Clonality: Monoclonal
Clone: 8B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR